Trimethylamin-N-oxid Dihydrat

Lieferant: Molekula

Synonyme: Trimethylamine oxide dihydrate

M23966397
MOLEM23966397EA 0 EUR
MOLEM23966397
Trimethylamin-N-oxid Dihydrat
Trimethylamin-N-oxid Dihydrat
Formel: C₃H₉NO·2H₂O
Molecular Weight: 111,14 g/mol (75,11 anhy)
MDL: MFCD00149077
CAS-Nummer: 62637-93-8
EINECS: 214-675-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR