D(+)-Tyrosin
Lieferant: Molekula
Synonyme:
(R)-(+)-p-Hydroxyphenylalanin, H-D-Tyr-OH, (R)-(+)-2-Amino-3-(4-hydroxyphenyl)propionsäure, D(+)-p-Hydroxyphenylalanin
M24028905
MOLEM24028905EA
0
EUR
MOLEM24028905
D(+)-Tyrosin
D(+)-Tyrosin
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol Siedepunkt: 351 °C (1013 hPa) Schmelzpunkt: 285 °C |
MDL:
MFCD00063073 CAS-Nummer: 556-02-5 EINECS: 209-112-6 Merck Index: 12,09970 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...