Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)

Lieferant: Molekula

Synonyme: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide

M24182077
MOLEM24182077EA 0 EUR
MOLEM24182077
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Formel: C₁₉H₁₅NO₅
Molecular Weight: 337,3 g/mol
MDL: MFCD00010733
CAS-Nummer: 82911-69-1
EINECS: 433-520-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR