Spermidin
Lieferant: Molekula
Synonyme:
N-(3-Aminopropyl)-1,4-diaminobutan, 4-Azaoctamethylendiamin
M24358880
MOLEM24358880EA
0
EUR
MOLEM24358880
Spermidin
Spermidin
Formel:
C₇H₁₉N₃ Molecular Weight: 145,25 g/mol Siedepunkt: 246 °C (1013 hPa) Schmelzpunkt: 20…25 °C Dichte: 0,925 g/cm³ (20 °C) |
MDL:
MFCD00008229 CAS-Nummer: 124-20-9 EINECS: 204-689-0 Merck Index: 12,08893 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...