Iminodibenzyl

Lieferant: Molekula

Synonyme: 10,11-Dihydro-5H-dibenzo[b,f]azepine

M24565519
MOLEM24565519EA 0 EUR
MOLEM24565519
Iminodibenzyl
Iminodibenzyl
Formel: C₁₄H₁₃N
Molecular Weight: 195,27 g/mol
MDL: MFCD00005070
CAS-Nummer: 494-19-9
EINECS: 207-787-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR