3-Indolcarbaldehyd
Lieferant: Molekula
Synonyme:
Indol-3-carboxaldehyd
M24811401
MOLEM24811401EA
0
EUR
MOLEM24811401
3-Indolcarbaldehyd
3-Indolcarbaldehyd
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 192…195 °C Dichte: 1,34 g/cm³ (20 °C) |
MDL:
MFCD00005622 CAS-Nummer: 487-89-8 EINECS: 207-665-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...