N(α)-Boc-L-tryptophan

Lieferant: Molekula

Synonyme: Boc-Trp-OH

M25104502
MOLEM25104502EA 0 EUR
MOLEM25104502
N(α)-Boc-L-tryptophan
N(α)-Boc-L-tryptophan
Formel: C₁₆H₂₀N₂O₄
Molecular Weight: 304,3 g/mol
MDL: MFCD00065595
CAS-Nummer: 13139-14-5
EINECS: 236-072-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR