2-Brombenzylalkohol
Lieferant: Molekula
Synonyme:
(2-Bromophenyl)methanol
M25429515
MOLEM25429515EA
0
EUR
MOLEM25429515
2-Brombenzylalkohol
2-Brombenzylalkohol
Formel:
C₇H₇BrO Molecular Weight: 187,04 g/mol Siedepunkt: 255 °C (1013 hPa) Schmelzpunkt: 80…82 °C |
MDL:
MFCD00004600 CAS-Nummer: 18982-54-2 EINECS: 242-719-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...