L(+)-Penicillamin

Lieferant: Molekula

Synonyme: (R)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine

M25587231
MOLEM25587231EA 0 EUR
MOLEM25587231
L(+)-Penicillamin
L(+)-Penicillamin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149,21 g/mol
MDL: MFCD00064303
CAS-Nummer: 1113-41-3
EINECS: 214-203-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR