(S)-(+)-β-Aminophenethylalkohol

Lieferant: Molekula

Synonyme: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

M25963926
MOLEM25963926EA 0 EUR
MOLEM25963926
(S)-(+)-β-Aminophenethylalkohol
(S)-(+)-β-Aminophenethylalkohol
Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
MDL: MFCD00064404
CAS-Nummer: 20989-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR