4-Fluorphenylhydraziniumchlorid
Lieferant: Molekula
Synonyme:
4-Fluorphenylhydrazin Hydrochlorid, 1-(4-Fluorphenyl)hydrazin Hydrochlorid, 4-Fluorphenylhydrazinhydrochlorid
M26460716
MOLEM26460716EA
0
EUR
MOLEM26460716
4-Fluorphenylhydraziniumchlorid
4-Fluorphenylhydraziniumchlorid
Formel:
C₆H₈ClFN₂ Molecular Weight: 162,59 g/mol |
MDL:
MFCD00012942 CAS-Nummer: 823-85-8 EINECS: 212-521-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...