Digitonin

Lieferant: Molekula

Synonyme: Digitonin

M26630625
MOLEM26630625EA 0 EUR
MOLEM26630625
Digitonin
Digitonin
Formel: C₅₆H₉₂O₂₉
Molecular Weight: 1229.312 g/mol
MDL: MFCD00077729
CAS-Nummer: 11024-24-1
EINECS: 234-255-6
UN: 3462
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR