Tetraethylammoniumbromid

Lieferant: Molekula

Synonyme: Tetrylammoniumbromid

M26739038
MOLEM26739038EA 0 EUR
MOLEM26739038
Tetraethylammoniumbromid
Tetraethylammoniumbromid
Formel: C₈H₂₀BrN
Schmelzpunkt: > 285 °C
Dichte: 1,43 g/cm³ (20 °C)
MDL: MFCD00011825
CAS-Nummer: 71-91-0
EINECS: 200-769-4
Merck Index: 13,09274

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR