3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat

Lieferant: Molekula

Synonyme: Dihydrochloride TMB, TMB-HCl

M27169695
MOLEM27169695EA 0 EUR
MOLEM27169695
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
Formel: C₁₆H₂₀N₂·2HCl·xH₂O
MDL: MFCD00150104
CAS-Nummer: 207738-08-7
EINECS: 264-769-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR