7-Methoxy-1-tetralon

Lieferant: Molekula

Synonyme: 7-Methoxy-3,4-dihydronaphthalen-1(2H)-one

M27521042
MOLEM27521042EA 0 EUR
MOLEM27521042
7-Methoxy-1-tetralon
7-Methoxy-1-tetralon
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,22 g/mol
MDL: MFCD00001696
CAS-Nummer: 6836-19-7
EINECS: 229-916-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR