2,3,4-Trimethoxybenzaldehyd
Lieferant: Molekula
M27948154
MOLEM27948154EA
0
EUR
MOLEM27948154
2,3,4-Trimethoxybenzaldehyd
2,3,4-Trimethoxybenzaldehyde
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196,2 g/mol Siedepunkt: 312 °C (1013 hPa) Schmelzpunkt: 30 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003310 CAS-Nummer: 2103-57-3 EINECS: 218-271-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...