(1S)-(+)-Campher-10-sulfonsäure

Lieferant: Molekula

Synonyme: D(+)-Campher-10-sulfonsäure, (1S)-(+)-2-Oxobornan-10-sulfonsäure

M28017028
MOLEM28017028EA 0 EUR
MOLEM28017028
(1S)-(+)-Campher-10-sulfonsäure
(1S)-(+)-Campher-10-sulfonsäure
Formel: C₁₀H₁₆O₄S
Molecular Weight: 232,3 g/mol
Siedepunkt: 375 °C (1013 hPa)
Schmelzpunkt: 195 °C
MDL: MFCD00064157
CAS-Nummer: 3144-16-9
EINECS: 221-554-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR