3-Methoxybenzylamin
Lieferant: Molekula
|
Achtung
|
Synonyme:
m-Anisylamine, 3-(Aminomethyl)anisole
M28782994
MOLEM28782994EA
0
EUR
MOLEM28782994
3-Methoxybenzylamin
3-Methoxybenzylamin
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol Siedepunkt: 238 °C (1013 hPa) Dichte: 1,07 g/cm³ (20 °C) |
MDL:
MFCD00008115 CAS-Nummer: 5071-96-5 EINECS: 225-779-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...