Benzophenonimin

Lieferant: Molekula

Synonyme: Benzophenoneimine

M28942128
MOLEM28942128EA 0 EUR
MOLEM28942128
Benzophenonimin
Benzophenonimin
Formel: C₁₃H₁₁N
Molecular Weight: 181.24 g/mol
MDL: MFCD00001760
CAS-Nummer: 1013-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR