2,3,4,5-Tetrafluorbenzoesäure
Lieferant: Molekula
|
Achtung
|
M29342429
MOLEM29342429EA
0
EUR
MOLEM29342429
2,3,4,5-Tetrafluorbenzoesäure
2,3,4,5-Tetrafluorobenzoic acid
Formel:
C₇H₂F₄O₂ Molecular Weight: 194,09 g/mol Siedepunkt: 135…137 °C (23 mmHg) Schmelzpunkt: 92…92,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009613 CAS-Nummer: 1201-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...