3-Phenoxypropionsäure

Lieferant: Molekula

Synonyme: 3-Penoxypropionic acid, 3-Phenoxypropanoic acid

M29353362
MOLEM29353362EA 0 EUR
MOLEM29353362
3-Phenoxypropionsäure
3-Phenoxypropionsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Siedepunkt: 235 °C (1013 hPa)
Schmelzpunkt: 92 °C
MDL: MFCD00002769
CAS-Nummer: 7170-38-9
EINECS: 230-518-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR