Pyridiniumchlorochromat

Lieferant: Molekula

Synonyme: PCC

M29524115
MOLEM29524115EA 0 EUR
MOLEM29524115
Pyridiniumchlorochromat
Pyridinium chlorochromate
Formel: C₅H₆ClCrNO₃
Molecular Weight: 215,56 g/mol
Schmelzpunkt: 205 °C
MDL: MFCD00013106
CAS-Nummer: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08064

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR