4-Chlorzimtsäure
Lieferant: Molekula
Synonyme:
4-Chlorcinnamsäure, 3-(p-Chlorphenyl)propensäure, 3-(4-Chlorphenyl)propensäure
M29907936
MOLEM29907936EA
0
EUR
MOLEM29907936
4-Chlorzimtsäure
4-Chlorzimtsäure
Formel:
C₉H₇ClO₂ Molecular Weight: 182,61 g/mol Siedepunkt: 311 °C (1013 hPa) Schmelzpunkt: 252…254 °C |
MDL:
MFCD00004396 CAS-Nummer: 1615-02-7 EINECS: 216-564-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...