5'-Brom-2'-hydroxyacetophenon

Lieferant: Molekula

Synonyme: 1-(5-Brom-2-hydroxyphenyl)ethan-1-on, 2-Acetyl-4-bromphenol

M30294608
MOLEM30294608EA 0 EUR
MOLEM30294608
5'-Brom-2'-hydroxyacetophenon
5'-Brom-2'-hydroxyacetophenon
Formel: C₈H₇BrO₂
MDL: MFCD00191850
CAS-Nummer: 1450-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR