3-Pyridinsulfonsäure

Lieferant: Molekula

M30531969
MOLEM30531969EA 0 EUR
MOLEM30531969
3-Pyridinsulfonsäure
3-Pyridinesulphonic acid
Formel: C₅H₅NO₃S
MDL: MFCD00006381
CAS-Nummer: 636-73-7
EINECS: 211-265-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR