Methyl-4-amino-2-methoxybenzoat
Lieferant: Molekula
Synonyme:
4-Amino-2-methoxybenzoesäuremethylester
M30571720
MOLEM30571720EA
0
EUR
MOLEM30571720
Methyl-4-amino-2-methoxybenzoat
Methyl-4-amino-2-methoxybenzoat
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol |
MDL:
MFCD00017202 CAS-Nummer: 27492-84-8 EINECS: 248-494-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...