4-(Methylamino)benzoesäure

Lieferant: Molekula

M30597436
MOLEM30597436EA 0 EUR
MOLEM30597436
4-(Methylamino)benzoesäure
4-(Methylamino)benzoic acid
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
Siedepunkt: 296 °C (1013 hPa)
Schmelzpunkt: 159 °C
MDL: MFCD00002535
CAS-Nummer: 10541-83-0
EINECS: 234-121-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR