Riboflavin-5'-phosphat Natriumsalz Dihydrat

Lieferant: Molekula

Synonyme: Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogenphosphate dihydrate

M31244850
MOLEM31244850EA 0 EUR
MOLEM31244850
Riboflavin-5'-phosphat Natriumsalz Dihydrat
Riboflavin-5'-phosphat Natriumsalz Dihydrat
Formel: C₁₇H₂₀N₄NaO₉P·2H₂O
Molecular Weight: 514.36 g/mol (478.34 anhy)
Lagertemperatur: Gefrierschrank
MDL: MFCD00150993
CAS-Nummer: 6184-17-4
EINECS: 204-988-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR