(S)-(-)-α,α-Diphenylprolinol

Lieferant: Molekula

Synonyme: (S)-(-)-α,α-Diphenyl-2-pyrrolidinemethanol

M31262339
MOLEM31262339EA 0 EUR
MOLEM31262339
(S)-(-)-α,α-Diphenylprolinol
(S)-(-)-α,α-Diphenylprolinol
Formel: C₁₇H₁₉NO
Molecular Weight: 253,35 g/mol
MDL: MFCD00075506
CAS-Nummer: 112068-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR