4-Amino-2-chlor-6,7-dimethoxychinazolin

Lieferant: Molekula

Synonyme: Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline

M31299379
MOLEM31299379EA 0 EUR
MOLEM31299379
4-Amino-2-chlor-6,7-dimethoxychinazolin
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Formel: C₁₀H₁₀ClN₃O₂
Molecular Weight: 239,66 g/mol
MDL: MFCD00051734
CAS-Nummer: 23680-84-4
EINECS: 245-821-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR