Cytidin

Lieferant: Molekula

Synonyme: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

M31533552
MOLEM31533552EA 0 EUR
MOLEM31533552
Cytidin
Cytidin
Formel: C₉H₁₃N₃O₅
Molecular Weight: 243,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006545
CAS-Nummer: 65-46-3
EINECS: 200-610-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR