Guanosin-5'-triphosphat Dinatriumsalz Dihydrat

Lieferant: Molekula

Synonyme: GTP disodium salt dihydrate

M32121500
MOLEM32121500EA 0 EUR
MOLEM32121500
Guanosin-5'-triphosphat Dinatriumsalz Dihydrat
Guanosin-5'-triphosphat Dinatriumsalz Dihydrat
Formel: C₁₀H₁₄N₅Na₂O₁₄P₃·2H₂O
Molecular Weight: 567,147 g/mol (anhy)
MDL: MFCD03410297
CAS-Nummer: 56001-37-7
EINECS: 259-940-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR