3,5-Diamino-1H-1,2,4-triazol

Lieferant: Molekula

Synonyme: Guanazol

M32317666
MOLEM32317666EA 0 EUR
MOLEM32317666
3,5-Diamino-1H-1,2,4-triazol
3,5-Diamino-1H-1,2,4-triazol
Formel: C₂H₅N₅
Molecular Weight: 99.1 g/mol
Siedepunkt: 425
Schmelzpunkt: 205
MDL: MFCD00005233
CAS-Nummer: 1455-77-2
EINECS: 215-937-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR