cis-4-Hydroxy-D(+)-prolin

Lieferant: Molekula

M32560054
MOLEM32560054EA 0 EUR
MOLEM32560054
cis-4-Hydroxy-D(+)-prolin
cis-4-Hydroxy-D(+)-prolin
Formel: C₅H₉NO₃
MDL: MFCD00005252
CAS-Nummer: 2584-71-6
EINECS: 219-963-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR