3-Chlorthiophen

Lieferant: Molekula

M32585806
MOLEM32585806EA 0 EUR
MOLEM32585806
3-Chlorthiophen
3-Chlorthiophen
Formel: C₄H₃ClS
Molecular Weight: 118,59 g/mol
Siedepunkt: 133…135 °C (1013 hPa)
Dichte: 1,276 g/cm³ (20 °C)
MDL: MFCD00043887
CAS-Nummer: 17249-80-8
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR