2-Acetylpyrrol
Lieferant: Molekula
Synonyme:
1-(1H-Pyrrol-2-yl)ethan-1-on, 1-(1H-Pyrrol-2-yl)-1-ethanon, 2-Acetyl-1H-pyrrol
M33539603
MOLEM33539603EA
0
EUR
MOLEM33539603
2-Acetylpyrrol
2-Acetylpyrrol
Formel:
C₆H₇NO Molecular Weight: 109,13 g/mol Siedepunkt: 220 °C (1013 hPa) Schmelzpunkt: 63…65 °C |
MDL:
MFCD00005220 CAS-Nummer: 1072-83-9 EINECS: 214-016-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...