4-Chlorpyridin-N-oxid

Lieferant: Molekula

M33671692
MOLEM33671692EA 0 EUR
MOLEM33671692
4-Chlorpyridin-N-oxid
4-Chlorpyridin-N-oxid
Formel: C₅H₄ClNO
MDL: MFCD00047425
CAS-Nummer: 1121-76-2
EINECS: 214-336-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR