5-Bromvanillin
Lieferant: Molekula
|
Achtung
|
Synonyme:
3-Brom-4-hydroxy-5-methoxybenzaldehyd
M33715221
MOLEM33715221EA
0
EUR
MOLEM33715221
5-Bromvanillin
5-Bromvanillin
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol Siedepunkt: 314 °C (1013 hPa) Schmelzpunkt: 166…167 °C |
MDL:
MFCD00006940 CAS-Nummer: 2973-76-4 EINECS: 221-016-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...