4-Brom-1-naphthylamin

Lieferant: Molekula

Synonyme: 1-Amino-4-bromnaphthalin

M34087347
MOLEM34087347EA 0 EUR
MOLEM34087347
4-Brom-1-naphthylamin
4-Brom-1-naphthylamin
Formel: C₁₀H₈BrN
Molecular Weight: 222,08 g/mol
MDL: MFCD00004023
CAS-Nummer: 2298-07-9
EINECS: 218-944-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR