(R)-(-)-1,2-Propandiol

Lieferant: Molekula

M34102771
MOLEM34102771EA 0 EUR
MOLEM34102771
(R)-(-)-1,2-Propandiol
(R)-(-)-1,2-Propandiol
Formel: C₃H₈O₂
MDL: MFCD00066248
CAS-Nummer: 4254-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR