L-Norvalin

Lieferant: Molekula

Synonyme: (S)-(+)-Norvalin, (S)-(+)-2-Aminovaleriansäure, (S)-(+)-2-Aminopentansäure, H-Nva-OH

M34368263
MOLEM34368263EA 0 EUR
MOLEM34368263
L-Norvalin
L-Norvalin
Formel: C₅H₁₁NO₂
Molecular Weight: 117,15 g/mol
Siedepunkt: 228 °C (1013 hPa)
Schmelzpunkt: 101 °C
MDL: MFCD00064421
CAS-Nummer: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR