2-(4-Fluorphenyl)indol

Lieferant: Molekula

M34436720
MOLEM34436720EA 0 EUR
MOLEM34436720
2-(4-Fluorphenyl)indol
2-(4-Fluorophenyl)indole
Formel: C₁₄H₁₀FN
MDL: MFCD00068178
CAS-Nummer: 782-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR