Methyl-6-brom-2-naphthoat

Lieferant: Molekula

Synonyme: Methyl 6-bromo-2-naphthoate

M34783101
MOLEM34783101EA 0 EUR
MOLEM34783101
Methyl-6-brom-2-naphthoat
Methyl-6-brom-2-naphthoat
Formel: C₁₂H₉BrO₂
Molecular Weight: 265,11 g/mol
MDL: MFCD00100408
CAS-Nummer: 33626-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR