Benzylchlorformiat

Lieferant: Molekula
Gefahr

Synonyme: CBZ-Cl (Z-Cl)

M34819962
MOLEM34819962EA 0 EUR
MOLEM34819962
Benzylchlorformiat
Benzylchlorformiat
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
MDL: MFCD00000640
CAS-Nummer: 501-53-1
EINECS: 207-925-0
UN: 1739
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR