Triclosan

Lieferant: Molekula

Synonyme: 5-Chloro-2-(2,4-dichlorophenoxy)phenol, 5-Chloro-2-(2, 4-dichlorophenoxy) phenol

M35047938
MOLEM35047938EA 0 EUR
MOLEM35047938
Triclosan
Triclosan
Formel: C₁₂H₇Cl₃O₂
Molecular Weight: 289,54 g/mol
MDL: MFCD00800992
CAS-Nummer: 3380-34-5
EINECS: 222-182-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR