β-Jonon

Lieferant: Molekula

M35676152
MOLEM35676152EA 0 EUR
MOLEM35676152
β-Jonon
β-Jonon
Formel: C₁₃H₂₀O
MDL: MFCD00001549
CAS-Nummer: 79-77-6
EINECS: 201-224-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR