Allyltrimethylsilan

Lieferant: Molekula

M36010201
MOLEM36010201EA 0 EUR
MOLEM36010201
Allyltrimethylsilan
Allyltrimethylsilan
Formel: H₂CCHCH₂Si(CH₃)₃
Siedepunkt: 84 °C (1013 hPa)
Dichte: 0,7139 g/cm³ (20 °C)
MDL: MFCD00008635
CAS-Nummer: 762-72-1
EINECS: 212-104-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR