(±)-trans-1,2-Diaminocyclohexan
Lieferant: Molekula
Synonyme:
(±)-trans-1,2-Cyclohexandiamin
M36339634
MOLEM36339634EA
0
EUR
MOLEM36339634
(±)-trans-1,2-Diaminocyclohexan
(±)-trans-1,2-Diaminocyclohexan
Formel:
C₆H₁₄N₂ Molecular Weight: 114,19 g/mol Siedepunkt: 187 °C (1013 hPa) Schmelzpunkt: 14,8 °C Dichte: 0,951 g/cm³ (20 °C) |
MDL:
MFCD00063747 CAS-Nummer: 1121-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...