Aminoacetaldehyddimethylacetal

Lieferant: Molekula

Synonyme: 2,2-Dimethoxyethanamine

M36385342
MOLEM36385342EA 0 EUR
MOLEM36385342
Aminoacetaldehyddimethylacetal
Aminoacetaldehyddimethylacetal
Formel: C₄H₁₁NO₂
Molecular Weight: 105,14 g/mol
MDL: MFCD00008135
CAS-Nummer: 22483-09-6
EINECS: 245-026-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR