4-Sulfamoylbenzoesäure

Lieferant: Molekula

Synonyme: 4-Sulphamoylbenzoic acid

M36458848
MOLEM36458848EA 0 EUR
MOLEM36458848
4-Sulfamoylbenzoesäure
4-Sulfamoylbenzoesäure
Formel: C₇H₇NO₄S
Molecular Weight: 201,2 g/mol
MDL: MFCD00007938
CAS-Nummer: 138-41-0
EINECS: 205-327-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR